missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,2-Dichloro-3-iodobenzene, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC222011000

 View more versions of this product

Catalog No. AC222011000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥96.0%
272.9
White
96.0
Glass bottle
97%
1, 2-Dichloro-3-iodobenzene
MFCD00001034
C6H3Cl2I
Crystalline Mass
100g
33° to 37°C
100.0
Authentic
97%
Cl2C6H3I
2401-21-0

Chemical Identifiers

2401-21-0
100.0
C6H3Cl2I
272.9
1, 2-Dichloro-3-iodobenzene, 97%
96.0
97%
Cl2C6H3I
MFCD00001034
≥96.0%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 219-276-0

TSCA : TSCA