missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,2-Dichloro-3-iodobenzene, 97%, ACROS Organics™

Chemical Identifiers

CAS 2401-21-0
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C6H3Cl2I
Linear Formula Cl2C6H3I
Formula Weight 272.9
MDL Number MFCD00001034
Chemical Name or Material 1, 2-Dichloro-3-iodobenzene, 97%
Assay ≥96.0%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC222011000 SDS Thermo Fisher Scientific
AC222011000
Glass bottle 100g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

2401-21-0
100.0
C6H3Cl2I
272.9
1, 2-Dichloro-3-iodobenzene, 97%
96.0
97%
Cl2C6H3I
MFCD00001034
≥96.0%

Specifications

Crystalline Mass
100g
33° to 37°C
100.0
Authentic
Cl2C6H3I
2401-21-0
1, 2-Dichloro-3-iodobenzene, 97%
272.9
White
96.0
Glass bottle
97%
MFCD00001034
C6H3Cl2I
≥96.0%