Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,2-Dibromotetrafluorobenzene, 99%

$88.36 - $297.08

Chemical Identifiers

Chemical Name or Material 1,2-Dibromotetrafluorobenzene
CAS 827-08-7
Molecular Formula C6Br2F4
Formula Weight 307.87
MDL Number MFCD00039209
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1662003

 
alfa aesar™
A1662003
1g Each for $88.36

AAA1662006

 
alfa aesar™
A1662006
5g Each for $297.08
Description & Specifications

Specifications

Formula Weight 307.87
Chemical Name or Material 1,2-Dibromotetrafluorobenzene
Boiling Point 197° to 199°C
Assay 99%
Quantity 1g
Flash Point 99°C (210°F)
Molecular Formula C6Br2F4
MDL Number MFCD00039209
CAS 827-08-7
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
1,2-Dibromotetrafluorobenzene is used as medicine and liquid crystal material intermediate. It helps in the synthesis of diphenyl(o-bromotetrafluorophenyl)phosphine and 2,3,4,5tetrafluorothioanisole.

Solubility
Insoluble in water.

Notes
Store in a cool, dry, conditions in well sealed container. Store away from oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.