Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,2-Dibromopropane, 98%

Chemical Identifiers

Chemical Name or Material 1,2-Dibromopropane
CAS 78-75-1
Molecular Formula C3H6Br2
Formula Weight 201.9
MDL Number MFCD00000148
UN Number UN1993
Beilstein 1718884
Merck Index 14,7853
Synonym Propylene dibromide; Propylene bromide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2299422

  SDS
alfa aesar™
B2299422
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 201.9
MDL Number MFCD00000148
Health Hazard 2 GHS H Statement
H330-H226-H302-H315-H319
Fatal if inhaled.
Flammable liquid and vapour.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
UN Number UN1993
Health Hazard 1 Danger
Melting Point −55°C
Molecular Formula C3H6Br2
Synonym Propylene dibromide; Propylene bromide
RTECS Number TX8574000
Quantity 100g
TSCA TSCA
Health Hazard 3 GHS P Statement
P210-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Beilstein 1718884
Merck Index 14,7853
Chemical Name or Material 1,2-Dibromopropane
Flash Point 35°C (95°F)
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Boiling Point 140° to 142°C
CAS 78-75-1
Assay 98%

Applications
1,2-Dibromopropane was used to study the effect of reductant concentration, reductant contact time and suspension pH on reductive dechlorination of carbon tetrachloride by soil manipulated with Fe(II) and HS-. It was used as internal standard during the determination of nitrogenous disinfection byproduct trichloronitromethane by gas chromatography mass spectrometry.

Solubility
Soluble in water (slightly), ether, and alcohol.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P210-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H330-H226-H302-H315-H319
Fatal if inhaled.
Flammable liquid and vapour.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.