Please login to your online account to display your discounted pricing

Alfa Aesar™ (1,2-Dibromoethyl)benzene, 97%

Chemical Identifiers

Chemical Name or Material (1,2-Dibromoethyl)benzene
CAS 93-52-7
Molecular Formula C8H8Br2
Formula Weight 263.96
MDL Number MFCD00000138
UN Number UN3261
Synonym 1,2-Dibromo-1-phenylethane; Styrene dibromide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2499014

  SDS
alfa aesar™
B2499014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2499022

  SDS
alfa aesar™
B2499022
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2499036

  SDS
alfa aesar™
B2499036
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 263.96
Chemical Name or Material (1,2-Dibromoethyl)benzene
TSCA TSCA
Melting Point 70° to 74°C
Boiling Point 139° to 140°C (15mmHg)
Assay 97%
Quantity 25g
Molecular Formula C8H8Br2
MDL Number MFCD00000138
CAS 93-52-7
UN Number UN3261
Synonym 1,2-Dibromo-1-phenylethane; Styrene dibromide
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.