Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,2-Dibromoethane-d4, 99%(Isotopic)

Chemical Identifiers

Chemical Name or Material 1,2-Dibromoethane-d4
CAS 22581-63-1
Name Note Isotopic
Formula Weight 191.89
MDL Number MFCD00037499
UN Number UN1605
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4227906

  SDS
alfa aesar™
4227906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 191.89
Chemical Name or Material 1,2-Dibromoethane-d4
Name Note Isotopic
UN Number UN1605
MDL Number MFCD00037499
Density 2.20g/mL
Refractive Index 1.5360
DOT Information Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: ETHYLENE DIBROMIDE
Boiling Point 132°C
Health Hazard 3 GHS P Statement
P273-P201-P309-P310
Avoid release to the environment.
Obtain special instructions before use.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Physical Form Liquid
Quantity 5g
Health Hazard 2 GHS H Statement
H301-H311-H331-H350-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 99%
Odor Slightly sweetish
Health Hazard 1 Danger
Melting Point 9°C
CAS 22581-63-1
Safety and Handling

Danger

GHS P Statement
P273-P201-P309-P310
Avoid release to the environment.
Obtain special instructions before use.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H301-H311-H331-H350-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.