missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,2-Dibromoethane, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC112790100

 View more versions of this product

Catalog No. AC112790100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

1,2-Dibromoethane
>104°C
Plastic Drum
2.173
Colorless to Yellow
10L
98.5
98.5% min. (GC)
BrCH2CH2Br
01,374; 04,126; 05,187; 12,570
1,2-dibromaethan, 1,2-dibroomethaan, 1,2-ethylene dibromide, aadibroom, alpha,beta-dibromoethane, bromuro di etile, ethane, 1,2-dibromo, ethylene bromide, ethylene dibromide, sym-dibromoethane
PAAZPARNPHGIKF-UHFFFAOYSA-N
1,2-dibromoethane
7839
187.86
99%
2.1730g/mL
Authentic
1.5375 to 1.5395
131.0°C to 132.0°C
9.0°C to 10.0°C
106-93-4
100.0
C2H4Br2
01,9
15,385
Solubility in water: 4g/L (20°C). Other solubilities: miscible with most organic solvents
C(CBr)Br
187.86
CHEBI:28534
Liquid

Chemical Identifiers

106-93-4
187.86
1,2-dibromaethan, 1,2-dibroomethaan, 1,2-ethylene dibromide, aadibroom, alpha,beta-dibromoethane, bromuro di etile, ethane, 1,2-dibromo, ethylene bromide, ethylene dibromide, sym-dibromoethane
CHEBI:28534
C(CBr)Br
C2H4Br2
PAAZPARNPHGIKF-UHFFFAOYSA-N
7839
1,2-dibromoethane
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause cancer.
Toxic to aquatic life with long lasting effects.
Toxic in contact with skin.
Toxic if inhaled.
May cause respiratory irritation.
Toxic if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF exposed or concerned: Get medical advice/attention.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 203-444-5

SDS Documents