Please login to your online account to display your discounted pricing

1,2-Dibromocyclopentene, 97%, Acros Organics


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC399420050

 View more versions of this product

Catalog No. AC399420050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material 1, 2-Dibromocyclopentene
Quantity 5g
CAS 75415-78-0
Packaging Glass bottle
Assay Percent Range 97%
Molecular Formula C5H6Br2
Formula Weight 225.9

Specifications

Chemical Name or Material 1, 2-Dibromocyclopentene
Assay Percent Range 97%
Molecular Formula C5H6Br2
Formula Weight 225.9