Learn More
1,2-Dibromobutane, 98%, Thermo Scientific Chemicals
$69.33 - $911.88
Chemical Identifiers
CAS | 533-98-2 |
---|---|
Molecular Formula | C4H8Br2 |
Molecular Weight (g/mol) | 215.92 |
MDL Number | MFCD00000157 |
InChI Key | CZWSZZHGSNZRMW-UHFFFAOYNA-N |
Synonym | butane, 1,2-dibromo, .alpha.-butylene dibromide, alpha-butylene dibromide, 1,2-dibomobutane, 1,2 dibromobutane, butane,2-dibromo, 1,2-dibromo-butane, alpha-butylene bromide, 1,2-butylene bromide, .alpha.-butylene bromide |
PubChem CID | 10792 |
IUPAC Name | 1,2-dibromobutane |
SMILES | CCC(Br)CBr |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2035618
|
Thermo Scientific Chemicals
B2035618 |
50 g |
Each for $69.33
|
|
|||||
AAB2035630
|
Thermo Scientific Chemicals
B2035630 |
250 g |
Each for $262.58
|
|
|||||
AAB203560B
|
Thermo Scientific Chemicals
B203560B |
1000 g |
Each for $911.88
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
533-98-2 | |
215.92 | |
CZWSZZHGSNZRMW-UHFFFAOYNA-N | |
10792 | |
CCC(Br)CBr |
C4H8Br2 | |
MFCD00000157 | |
butane, 1,2-dibromo, .alpha.-butylene dibromide, alpha-butylene dibromide, 1,2-dibomobutane, 1,2 dibromobutane, butane,2-dibromo, 1,2-dibromo-butane, alpha-butylene bromide, 1,2-butylene bromide, .alpha.-butylene bromide | |
1,2-dibromobutane |
Specifications
533-98-2 | |
1.789 | |
>110°C (230°F) | |
1.5141 | |
50 g | |
14,1566 | |
CZWSZZHGSNZRMW-UHFFFAOYNA-N | |
1,2-dibromobutane | |
10792 | |
98% |
-65°C | |
166°C to 167°C | |
C4H8Br2 | |
MFCD00000157 | |
1731380 | |
butane, 1,2-dibromo, .alpha.-butylene dibromide, alpha-butylene dibromide, 1,2-dibomobutane, 1,2 dibromobutane, butane,2-dibromo, 1,2-dibromo-butane, alpha-butylene bromide, 1,2-butylene bromide, .alpha.-butylene bromide | |
CCC(Br)CBr | |
215.92 | |
215.93 | |
1,2-Dibromobutane |
Safety and Handling
GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 208-581-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only