Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,2-Dibromo-4,5-(methylenedioxy)benzene, 98%

$109.92 - $438.69

Chemical Identifiers

Chemical Name or Material 1,2-Dibromo-4,5-(methylenedioxy)benzene
CAS 5279-32-3
Molecular Formula C7H4Br2O2
Formula Weight 279.93
MDL Number MFCD00094476
Beilstein 145633
Synonym 5,6-Dibromo-1,3-benzodioxole; 4,5-Methylenedioxy-1,2-dibromobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1310909

  SDS
alfa aesar™
L1310909
10g Each for $109.92

AAL1310918

  SDS
alfa aesar™
L1310918
50g Each for $438.69
Description & Specifications

Specifications

Formula Weight 279.93
Chemical Name or Material 1,2-Dibromo-4,5-(methylenedioxy)benzene
Assay 98%
Molecular Formula C7H4Br2O2
MDL Number MFCD00094476
Beilstein 145633
Synonym 5,6-Dibromo-1,3-benzodioxole; 4,5-Methylenedioxy-1,2-dibromobenzene
Melting Point 83° to 85°C
Quantity 10g
CAS 5279-32-3