Please login to your online account to display your discounted pricing

1,2-Dibromo-3,5-difluorobenzene, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 1, 2-Dibromo-3, 5-difluorobenzene
Name Note 99%
CAS 139215-43-3
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C6 H2 Br2 F2
Linear Formula Br2C6H2F2
Formula Weight 271.88
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC249480250

  SDS
thermo fisher scientific
249480250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline Powder or Flakes
Assay ≥98.5%
Quantity 25g
Color White to off-white
TSCA TSCA
Molecular Formula C6 H2 Br2 F2
Chemical Name or Material 1, 2-Dibromo-3, 5-difluorobenzene
CAS 139215-43-3
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS Max % 100.0
CAS Min % 98.5
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula Br2C6H2F2
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
MDL Number MFCD00042559
Assay Percent Range 99%
Name Note 99%
Melting Point 36° to 39°C
Formula Weight 271.88