Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,2-Dibromo-3,5-difluorobenzene, 98%

$30.45 - $83.18

Chemical Identifiers

Chemical Name or Material 1,2-Dibromo-3,5-difluorobenzene
CAS 139215-43-3
Molecular Formula C6H2Br2F2
Formula Weight 271.89
MDL Number MFCD00042559
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2563906

  SDS
alfa aesar™
B2563906
5g Each for $30.45

AAB2563914

  SDS
alfa aesar™
B2563914
25g Each for $83.18
Description & Specifications

Specifications

Formula Weight 271.89
Chemical Name or Material 1,2-Dibromo-3,5-difluorobenzene
Melting Point 36° to 38°C
Assay 98%
Quantity 5g
Flash Point >110°C (230°F)
Molecular Formula C6H2Br2F2
MDL Number MFCD00042559
CAS 139215-43-3
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.