missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,2-Dibromo-2-methylpropane, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC406492500

 View more versions of this product

Catalog No. AC406492500

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

1, 2-Dibromo-2-methylpropane
Clear colorless
98%
Authentic
100.0
1.5083 to 1.5103 (20°C, 589nm)
≥97.5%
C4H8Br2
215.92
1, 2-Dibromoisobutane
1.7900g/mL
250g
594-34-3
97.5
Glass bottle
Liquid
98%
(CH3)2CBrCH2Br
MFCD00000126

Chemical Identifiers

594-34-3
100.0
C4H8Br2
215.92
1, 2-Dibromoisobutane
≥97.5%
97.5
98%
(CH3)2CBrCH2Br
MFCD00000126
1, 2-Dibromo-2-methylpropane, 98%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Suspected of causing cancer.

GHS P Statement:
Use personal protective equipment as required.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 209-835-7

RTECSNumber : TX8810000

TSCA : TSCA