missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific Chemicals 1,2-Dibromo-1,1,2-trichloroethane, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 279030050
Specifications
1,2-Dibromo-1,1,2-trichloroethane | |
13749-38-7 | |
100.0 | |
>200°C | |
98.5% min. (GC) | |
C2HBr2Cl3 | |
MFCD00018841 | |
ethane,1,2-dibromo-1,1,2-trichloro, acmc-20ap6p, 1,2-dibromotrichloro ethane | |
C(C(Cl)(Cl)Br)(Cl)Br | |
291.2 | |
291.2 | |
Liquid |
99% | |
98.5 | |
Colorless | |
Authentic | |
Glass bottle | |
1.5700 to 1.5720 | |
5 g | |
VXKOVUFOVCJHBA-UHFFFAOYSA-N | |
1,2-dibromo-1,1,2-trichloroethane | |
114562 | |
99% |
Chemical Identifiers
13749-38-7 | |
291.2 | |
VXKOVUFOVCJHBA-UHFFFAOYSA-N | |
114562 | |
C(C(Cl)(Cl)Br)(Cl)Br |
C2HBr2Cl3 | |
MFCD00018841 | |
ethane,1,2-dibromo-1,1,2-trichloro, acmc-20ap6p, 1,2-dibromotrichloro ethane | |
1,2-dibromo-1,1,2-trichloroethane |
Safety and Handling
EINECSNumber : 237-330-1
RUO â Research Use Only