missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1,2-Dibenzoylbenzene, 97%, Thermo Scientific Chemicals
$70.95 - $177.36
Chemical Identifiers
CAS | 1159-86-0 |
---|---|
Molecular Formula | C20H14O2 |
Molecular Weight (g/mol) | 286.33 |
MDL Number | MFCD00003078 |
InChI Key | OJLABXSUFRIXFL-UHFFFAOYSA-N |
Synonym | 1,2-dibenzoylbenzene, 1,2-phenylenebis phenylmethanone, 2-benzoylbenzophenone, 2-benzoylphenyl phenyl methanone, methanone, 1,2-phenylenebis phenyl, ortho-dibenzoylbenzene, o-dibenzoylbenzene, methanone,1,1'-1,2-phenylene bis 1-phenyl, phenyl 2-phenylcarbonyl phenyl ketone |
PubChem CID | 70875 |
IUPAC Name | (2-benzoylphenyl)-phenylmethanone |
SMILES | C1=CC=C(C=C1)C(=O)C2=CC=CC=C2C(=O)C3=CC=CC=C3 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL01592MD
|
Thermo Scientific Chemicals
L01592MD |
250 mg |
Each for $70.95
|
|
|||||
AAL0159203
|
Thermo Scientific Chemicals
L0159203 |
1 g |
Each for $177.36
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1159-86-0 | |
286.33 | |
OJLABXSUFRIXFL-UHFFFAOYSA-N | |
70875 | |
C1=CC=C(C=C1)C(=O)C2=CC=CC=C2C(=O)C3=CC=CC=C3 |
C20H14O2 | |
MFCD00003078 | |
1,2-dibenzoylbenzene, 1,2-phenylenebis phenylmethanone, 2-benzoylbenzophenone, 2-benzoylphenyl phenyl methanone, methanone, 1,2-phenylenebis phenyl, ortho-dibenzoylbenzene, o-dibenzoylbenzene, methanone,1,1'-1,2-phenylene bis 1-phenyl, phenyl 2-phenylcarbonyl phenyl ketone | |
(2-benzoylphenyl)-phenylmethanone |
Specifications
1159-86-0 | |
C20H14O2 | |
250 mg | |
1,2-dibenzoylbenzene, 1,2-phenylenebis phenylmethanone, 2-benzoylbenzophenone, 2-benzoylphenyl phenyl methanone, methanone, 1,2-phenylenebis phenyl, ortho-dibenzoylbenzene, o-dibenzoylbenzene, methanone,1,1'-1,2-phenylene bis 1-phenyl, phenyl 2-phenylcarbonyl phenyl ketone | |
C1=CC=C(C=C1)C(=O)C2=CC=CC=C2C(=O)C3=CC=CC=C3 | |
286.33 | |
286.33 | |
1,2-Dibenzoylbenzene |
145°C to 148°C | |
MFCD00003078 | |
789077 | |
OJLABXSUFRIXFL-UHFFFAOYSA-N | |
(2-benzoylphenyl)-phenylmethanone | |
70875 | |
97% |
Safety and Handling
EINECSNumber : 214-597-2
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only