Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,2-Dianilinoethane, 98+%

$37.22 - $361.62

Chemical Identifiers

Chemical Name or Material 1,2-Dianilinoethane
CAS 150-61-8
Molecular Formula C6H5NHCH2CH2NHC6H5
Formula Weight 212.3
MDL Number MFCD00003019
Beilstein 646740
Merck Index 14,2990
Synonym N,N'-Diphenylethylenediamine; Wanzlick's Reagent
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2309806

  SDS
alfa aesar™
B2309806
5g Each for $37.22

AAB2309814

  SDS
alfa aesar™
B2309814
25g Each for $103.97

AAB2309822

  SDS
alfa aesar™
B2309822
100g Each for $361.62
Description & Specifications

Specifications

Formula Weight 212.3
Beilstein 646740
Quantity 5g
TSCA TSCA
Boiling Point 228°C (12mmHg)
Molecular Formula C6H5NHCH2CH2NHC6H5
Chemical Name or Material 1,2-Dianilinoethane
Health Hazard 1 Danger
CAS 150-61-8
Health Hazard 2 GHS H Statement
H301-H311-H332
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
MDL Number MFCD00003019
Merck Index 14,2990
Synonym N,N'-Diphenylethylenediamine; Wanzlick's Reagent
Assay ≥98%
Melting Point 64° to 68°C
RTECS Number KV4800000
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
1,2-Dianilinoethane can forms crystalline imidazolidines with aldehydes in the presence of AcOH. N-donating ligand. Can be used as a trapping agent for aldehydes.

Solubility
Sparingly soluble in water 0.072 g/L @ 25°C.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H332
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.