Learn More
1,2-Dianilinoethane, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2309806
Description
1,2-Dianilinoethane can forms crystalline imidazolidines with aldehydes in the presence of AcOH. N-donating ligand. Can be used as a trapping agent for aldehydes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,2-Dianilinoethane can forms crystalline imidazolidines with aldehydes in the presence of AcOH. N-donating ligand. Can be used as a trapping agent for aldehydes.
Solubility
Sparingly soluble in water 0.072 g/L @ 25°C.
Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
Specifications
1,2-Dianilinoethane | |
64°C to 68°C | |
C14H16N2 | |
5 g | |
14,2990 | |
Sparingly soluble in water 0.072g/L @ 25°C. | |
C(CNC1=CC=CC=C1)NC1=CC=CC=C1 | |
212.30 | |
212.3 |
150-61-8 | |
228°C (12 mmHg) | |
MFCD00003019 | |
646740 | |
1,2-dianilinoethane, n,n'-diphenylethylenediamine, stabilite, n,n'-ethylenedianiline, nodx, n1,n2-diphenylethane-1,2-diamine, sym-diphenylethylenediamine, 1,2-ethanediamine, n,n'-diphenyl, aniline, n,n'-ethylenedi, n,n'-difenylethylendiamin | |
NOUUUQMKVOUUNR-UHFFFAOYSA-N | |
N,N'-diphenylethane-1,2-diamine | |
67422 | |
≥98% |
Chemical Identifiers
150-61-8 | |
212.30 | |
NOUUUQMKVOUUNR-UHFFFAOYSA-N | |
67422 | |
C(CNC1=CC=CC=C1)NC1=CC=CC=C1 |
C14H16N2 | |
MFCD00003019 | |
1,2-dianilinoethane, n,n'-diphenylethylenediamine, stabilite, n,n'-ethylenedianiline, nodx, n1,n2-diphenylethane-1,2-diamine, sym-diphenylethylenediamine, 1,2-ethanediamine, n,n'-diphenyl, aniline, n,n'-ethylenedi, n,n'-difenylethylendiamin | |
N,N'-diphenylethane-1,2-diamine |
Safety and Handling
GHS H Statement
H301-H311-H332
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
GHS P Statement
P261-P280-P301+P310-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
EINECSNumber : 205-765-6
RTECSNumber : KV4800000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only