Please login to your online account to display your discounted pricing

1,2-Diaminopropane-N,N,N',N'-tetraacetic acid, 98+%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  169660050

 View more versions of this product

Catalog No. AC169660050

  • / Each
 

Description & Specifications

Chemical Identifiers

Infrared Spectrum Authentic
Packaging Glass bottle
Sulfated Ash 0.2% max.
Color White
Quantity 5g
Chemical Name or Material 1,2-Diaminopropane-N,N,N',N'-tetraacetic acid
Name Note 98+%
CAS 4408-81-5
CAS Min % 98.0
CAS Max % 100.0
Physical Form White micro-crystalline powder
Assay 98.0% min.
Assay Percent Range 98+%
Molecular Formula C11H18N2O8
Linear Formula (HO2CCH2)2NCH(CH3)CH2N(CH2CO2H)2
Formula Weight 306.26g/mol
MDL Number MFCD00004286
Beilstein 04, III, 1190
Synonym (Propylenedinitrilo)tetraacetic acid
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
RTECS Number AJ3500000
EINECS Number 224-559-7
TSCA TSCA

Specifications

Chemical Name or Material 1,2-Diaminopropane-N,N,N',N'-tetraacetic acid
Name Note 98+%
CAS 4408-81-5
CAS Min % 98.0
CAS Max % 100.0
Assay 98.0% min.
Assay Percent Range 98+%
Molecular Formula C11H18N2O8
Linear Formula (HO2CCH2)2NCH(CH3)CH2N(CH2CO2H)2
Formula Weight 306.26g/mol
MDL Number MFCD00004286
Synonym (Propylenedinitrilo)tetraacetic acid

Chelation/Complexation reagent

Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

SDS & Safety Documents