Learn More
1,2-Diaminopropane, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 152400025
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
1, 2-Diaminopropane | |
98.5 | |
0.8700g/mL | |
33°C | |
Glass bottle | |
1.4450 to 1.4470 | |
MFCD00008089 | |
04,257 | |
15,7965 | |
Solubility in water: soluble. Other solubilities: insoluble in ether, soluble in acetone, benzene, chloroform and, ethanol | |
CC(N)CN | |
74.13 | |
CHEBI:30630 | |
99% |
78-90-0 | |
100.0 | |
119.0°C to 120.0°C | |
98.5% min. (GC) | |
C3H10N2 | |
CH3CH(NH2)CH2NH2 | |
2.5L | |
0.87 | |
1,2-diaminopropane, propylenediamine, 1,2-propanediamine, propylene diamine, 2,3-diaminopropane, 1,2-propylenediamine, 1,2 diaminopropane, ccris 4863, 1-methylethylenediamine, 2-aminopropylamine | |
AOHJOMMDDJHIJH-UHFFFAOYNA-N | |
propane-1,2-diamine | |
6567 | |
74.13 |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Toxic in contact with skin.
Flammable liquid and vapor.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cau
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth.
GHS Signal Word: Danger
EINECSNumber : 201-155-9
RUO â Research Use Only