missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,2-Diaminoanthraquinone, tech. 90%

Manufacturer:  Alfa Aesar™ L1174114

 View more versions of this product

Catalog No. AAL1174114



Specifications

Specifications

1,2-Diaminoanthraquinone
~288°C
1758-68-5
MFCD00001219
1,2-diaminoanthraquinone, 9,10-anthracenedione, 1,2-diamino, 1,2-daa, 1,2-daa russian, 1,2-diamino-9,10-anthraquinone, unii-h5p6yp29va, diaminoanthraquinone, anthraquinone, 1,2-diamino, ccris 5602, 1,2-diamino-9,10-anthracenedione
C1=CC=C2C(=C1)C(=O)C3=C(C2=O)C(=C(C=C3)N)N
238.246
238.25
Technical
Odorless
25g
C14H10N2O2
2125604
LRMDXTVKVHKWEK-UHFFFAOYSA-N
1,2-diaminoanthracene-9,10-dione
15652
90%

Chemical Identifiers

1758-68-5
238.246
LRMDXTVKVHKWEK-UHFFFAOYSA-N
15652
C1=CC=C2C(=C1)C(=O)C3=C(C2=O)C(=C(C=C3)N)N
C14H10N2O2
MFCD00001219
1,2-diaminoanthraquinone, 9,10-anthracenedione, 1,2-diamino, 1,2-daa, 1,2-daa russian, 1,2-diamino-9,10-anthraquinone, unii-h5p6yp29va, diaminoanthraquinone, anthraquinone, 1,2-diamino, ccris 5602, 1,2-diamino-9,10-anthracenedione
1,2-diaminoanthracene-9,10-dione
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

RTECSNumber : CB6200000

TSCA : TSCA

Documents