Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,2-Diaminoanthraquinone, tech. 90%


Quantity 5g

Manufacturer: alfa aesar™  L11741-06

 View more versions of this product

Catalog No. AAL1174106

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Odor Odorless
Quantity 5g
Melting Point ≈288°C
Quantity 5g
Chemical Name or Material 1,2-Diaminoanthraquinone
CAS 1758-68-5
Purity Grade Technical Grade
Assay 90%
Molecular Formula C14H10N2O2
Formula Weight 238.25g/mol
MDL Number MFCD00001219
Beilstein 2125604
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RTECS Number CB6200000
TSCA TSCA

Specifications

Chemical Name or Material 1,2-Diaminoanthraquinone
CAS 1758-68-5
Purity Grade Technical Grade
Molecular Formula C14H10N2O2
Formula Weight 238.25g/mol
MDL Number MFCD00001219
Beilstein 2125604
Accessories

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.