missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,2-Di(p-tolyl)ethane, 98%

$51.88 - $183.18

Chemical Identifiers

CAS 538-39-6
Molecular Formula C16H18
Formula Weight 210.32
MDL Number MFCD00026025
Beilstein 1909630
Merck Index 14,3380
Chemical Name or Material 1,2-Di(p-tolyl)ethane
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0887603 SDS Alfa Aesar™
L0887603
1g Each for $51.88
AAL0887606 SDS Alfa Aesar™
L0887606
5g Each for $183.18
Specifications Description & Specifications

Chemical Identifiers

538-39-6
210.32
1909630
1,2-Di(p-tolyl)ethane
C16H18
MFCD00026025
14,3380

Specifications

296° to 298°C (730mmHg)
1g
C16H18
MFCD00026025
14,3380
98%
80° to 82°C
538-39-6
210.32
1909630
1,2-Di(p-tolyl)ethane

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : YT4380000