Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(2-Di-n-propylaminoethyl)piperazine, 99%

$71.72 - $298.34

Chemical Identifiers

Chemical Name or Material 1-(2-Di-n-propylaminoethyl)piperazine
CAS 496808-01-6
Molecular Formula C12H27N3
Formula Weight 213.37
MDL Number MFCD00191215
Synonym N,N-Di-n-propyl-1-piperazineethylamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5068603

  SDS
alfa aesar™
H5068603
1g Each for $71.72

AAH5068606

  SDS
alfa aesar™
H5068606
5g Each for $298.34
Description & Specifications

Specifications

Formula Weight 213.37
Quantity 1g
Assay 99%
MDL Number MFCD00191215
CAS 496808-01-6
Molecular Formula C12H27N3
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 1-(2-Di-n-propylaminoethyl)piperazine
Synonym N,N-Di-n-propyl-1-piperazineethylamine
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.