missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,2-Di(3-indenyl)ethane, 98%

$37.62 - $118.61

Chemical Identifiers

CAS 18657-57-3
Molecular Formula C20H18
Molecular Weight (g/mol) 258.364
MDL Number MFCD00191814
InChI Key CQAQBIQKEFJNRZ-UHFFFAOYSA-N
Synonym 1,2-bis 3-indenyl ethane, 1,2-di 1h-inden-3-yl ethane, 1,2-di 3-indenyl ethane, 1-2-3h-inden-1-yl ethyl-3h-indene, 1?2-bis 3-indenyl ethane, 1h-indene, 3,3'-1,2-ethanediyl bis, 1h-indene,3,3'-1,2-ethanediyl bis, 3,3'-ethylenebis 1h-indene, 3-2-3h-inden-1-yl ethyl-1h-indene, acmc-209eoe
PubChem CID 3488033
IUPAC Name 3-[2-(3H-inden-1-yl)ethyl]-1H-indene
SMILES C1C=C(C2=CC=CC=C21)CCC3=CCC4=CC=CC=C43
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2719503
View Documents
Alfa Aesar™
H2719503
1g
Each for $37.62
 
AAH2719506
View Documents
Alfa Aesar™
H2719506
5g
Each for $118.61
 
Description

Description

Specifications

Chemical Identifiers

18657-57-3
258.364
CQAQBIQKEFJNRZ-UHFFFAOYSA-N
3488033
C1C=C(C2=CC=CC=C21)CCC3=CCC4=CC=CC=C43
C20H18
MFCD00191814
1,2-bis 3-indenyl ethane, 1,2-di 1h-inden-3-yl ethane, 1,2-di 3-indenyl ethane, 1-2-3h-inden-1-yl ethyl-3h-indene, 1?2-bis 3-indenyl ethane, 1h-indene, 3,3'-1,2-ethanediyl bis, 1h-indene,3,3'-1,2-ethanediyl bis, 3,3'-ethylenebis 1h-indene, 3-2-3h-inden-1-yl ethyl-1h-indene, acmc-209eoe
3-[2-(3H-inden-1-yl)ethyl]-1H-indene

Specifications

121°C to 125°C
18657-57-3
MFCD00191814
CQAQBIQKEFJNRZ-UHFFFAOYSA-N
3-[2-(3H-inden-1-yl)ethyl]-1H-indene
3488033
98%
1g
C20H18
1,2-bis 3-indenyl ethane, 1,2-di 1h-inden-3-yl ethane, 1,2-di 3-indenyl ethane, 1-2-3h-inden-1-yl ethyl-3h-indene, 1?2-bis 3-indenyl ethane, 1h-indene, 3,3'-1,2-ethanediyl bis, 1h-indene,3,3'-1,2-ethanediyl bis, 3,3'-ethylenebis 1h-indene, 3-2-3h-inden-1-yl ethyl-1h-indene, acmc-209eoe
C1C=C(C2=CC=CC=C21)CCC3=CCC4=CC=CC=C43
258.364
258.36
1,2-Di(3-indenyl)ethane
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS