Please login to your online account to display your discounted pricing

1,2-Decanediol, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 1, 2-Decanediol
CAS 1119-86-4
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C10H22O2
Linear Formula CH3(CH2)7CH(OH)CH2OH
Formula Weight 174.28
MDL Number MFCD00010739
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC372590100

  SDS
thermo fisher scientific
AC372590100
Glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC372590500

  SDS
thermo fisher scientific
AC372590500
Glass bottle 50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Powder
Formula Weight 174.28
Quantity 10g
Color White
Melting Point 47° to 51°C
CAS Max % 100.0
Packaging Glass bottle
Infrared Spectrum Authentic
Chemical Name or Material 1, 2-Decanediol
Assay Percent Range 98%
Molecular Formula C10H22O2
MDL Number MFCD00010739
CAS 1119-86-4
CAS Min % 97.5
Linear Formula CH3(CH2)7CH(OH)CH2OH