Please login to your online account to display your discounted pricing

1,2-Decanediol, 98%, ACROS Organics™


Quantity 10g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC372590100

 View more versions of this product

Catalog No. AC372590100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Powder
Formula Weight 174.28
Quantity 10g
Color White
Melting Point 47° to 51°C
CAS Max % 100.0
Packaging Glass bottle
Infrared Spectrum Authentic
Chemical Name or Material 1, 2-Decanediol
Assay Percent Range 98%
Molecular Formula C10H22O2
MDL Number MFCD00010739
CAS 1119-86-4
CAS Min % 97.5
Linear Formula CH3(CH2)7CH(OH)CH2OH

Specifications

Chemical Name or Material 1, 2-Decanediol
CAS 1119-86-4
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C10H22O2
Linear Formula CH3(CH2)7CH(OH)CH2OH
Formula Weight 174.28
MDL Number MFCD00010739