Learn More
1,2-Cyclohexanedione, 98+%
CAS: 765-87-7 | C6H8O2 | 112.128 g/mol
Supplier: Thermo Scientific Chemicals A1440114
Description
1,2-Cyclohexanedione is utilized as a substrate to study enzyme cyclohexane-1,2-dione hydrolase, which is a new tool to degrade alicyclic compounds. It also acts as a specific reagent for arginine residues.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,2-Cyclohexanedione is utilized as a substrate to study enzyme cyclohexane-1,2-dione hydrolase, which is a new tool to degrade alicyclic compounds. It also acts as a specific reagent for arginine residues.
Solubility
Soluble in water and alcohol.
Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place.
Specifications
| 1,2-Cyclohexanedione | |
| 35°C to 38°C | |
| 193°C to 195°C | |
| C6H8O2 | |
| MFCD00001648 | |
| 507419 | |
| Soluble in water and alcohol. | |
| C1CCC(=O)C(=O)C1 | |
| 112.128 | |
| CHEBI:41674 | |
| ≥98% |
| 765-87-7 | |
| 1.12 | |
| 84°C (183°F) | |
| 1.519 | |
| 25 g | |
| 1,2-cyclohexanedione, 1,2-dioxocyclohexane, cyclohexanedione, 1,2-cyclohexadione, cyclohexan-1,2-dione, ccris 6296, unii-75c1ovw0fj, 75c1ovw0fj, 1,2-cyclohexanedione,ketone form, dihydrocatechol | |
| OILAIQUEIWYQPH-UHFFFAOYSA-N | |
| cyclohexane-1,2-dione | |
| 13006 | |
| 112.13 |
Chemical Identifiers
| 765-87-7 | |
| MFCD00001648 | |
| 1,2-cyclohexanedione, 1,2-dioxocyclohexane, cyclohexanedione, 1,2-cyclohexadione, cyclohexan-1,2-dione, ccris 6296, unii-75c1ovw0fj, 75c1ovw0fj, 1,2-cyclohexanedione,ketone form, dihydrocatechol | |
| CHEBI:41674 |
| C6H8O2 | |
| OILAIQUEIWYQPH-UHFFFAOYSA-N | |
| 13006 | |
| cyclohexane-1,2-dione |
Safety and Handling
P264b-P270-P301+P312-P330-P501c
H302
EINECSNumber : 212-155-3
RTECSNumber : GV0340000
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only