missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,2-Cyclohexanediol, Mixture of cis and trans 98%, ACROS Organics™

Manufacturer:  Acros Organics 111130250

 View more versions of this product

Catalog No. AC111130250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

1, 2-Cyclohexanediol
Plastic bottle
73°C to 77°C
98%
97.5
98%
C6H10(OH)2
06, 740
PFURGBBHAOXLIO-UHFFFAOYSA-N
cyclohexane-1,2-diol
13601
116.16
≥97.5%
Authentic
White
25g
931-17-9
100.0
C6H12O2
MFCD00003861
1,2-cyclohexanediol, 2-hydroxycyclohexanol, pyrocatechitol, 1,2-dihydroxycyclohexane, brenzkatechin, 1,2-benzenediol, hexahydro, brenzkatechin german, 1,2-trans-cyclohexanediol, trans-1,2-dihydroxycyclohexane, 1,2-cyclohexanediol, mixture of cis and trans
C1CCC(C(C1)O)O
116.16
CHEBI:24567
Crystalline Powder

Chemical Identifiers

931-17-9
116.16
PFURGBBHAOXLIO-UHFFFAOYSA-N
13601
cyclohexane-1,2-diol
C6H12O2
MFCD00003861
1,2-cyclohexanediol, 2-hydroxycyclohexanol, pyrocatechitol, 1,2-dihydroxycyclohexane, brenzkatechin, 1,2-benzenediol, hexahydro, brenzkatechin german, 1,2-trans-cyclohexanediol, trans-1,2-dihydroxycyclohexane, 1,2-cyclohexanediol, mixture of cis and trans
CHEBI:24567
C1CCC(C(C1)O)O
Safety and Handling

Safety and Handling

EINECSNumber : 213-229-8

RTECSNumber : GV0220000

TSCA : TSCA

SDS Documents