missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,2-Cyclohexanedicarboxylic anhydride, cis + trans, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L2000136

 View more versions of this product

Catalog No. AAL2000136



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1,2-Cyclohexanedicarboxylic anhydride
85-42-7
158°C (17 mmHg)
C8H10O3
500 g
Moisture sensitive
MUTGBJKUEZFXGO-UHFFFAOYNA-N
3a,4,5,6,7,7a-hexahydro-2-benzofuran-1,3-dione
85689
154.17
Cis + Trans
32°C to 34°C
275°C (527°F)
MFCD00064863,MFCD00674195
83213
hexahydrophthalic anhydride, hexahydroisobenzofuran-1,3-dione, hhpa, 1,2-cyclohexanedicarboxylic anhydride, lekutherm hardener h, hexahydrophthalic acid anhydride, 1,3-isobenzofurandione, hexahydro, araldite ht 907, cyclohexane-1,2-dicarboxylic anhydride, 1,2-cyclohexanedicarboxylic acid anhydride
O=C1OC(=O)C2CCCCC12
154.17
CHEBI:103210
97%

Chemical Identifiers

85-42-7
154.17
MUTGBJKUEZFXGO-UHFFFAOYNA-N
85689
3a,4,5,6,7,7a-hexahydro-2-benzofuran-1,3-dione
C8H10O3
MFCD00064863,MFCD00674195
hexahydrophthalic anhydride, hexahydroisobenzofuran-1,3-dione, hhpa, 1,2-cyclohexanedicarboxylic anhydride, lekutherm hardener h, hexahydrophthalic acid anhydride, 1,3-isobenzofurandione, hexahydro, araldite ht 907, cyclohexane-1,2-dicarboxylic anhydride, 1,2-cyclohexanedicarboxylic acid anhydride
CHEBI:103210
O=C1OC(=O)C2CCCCC12
Safety and Handling

Safety and Handling

GHS H Statement
H334-H318-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes serious eye damage.
May cause an allergic skin reaction.

P261-P272-P280-P285-P302+P352-P304+P341-P305+P351+P338-P310-P333+P313-P342+P311-P363-P501c

H317-H318-H334

EINECSNumber : 201-604-9

RTECSNumber : NP6895168

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Argon

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only