Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(2-Cyanophenyl)piperazine, 97%

Chemical Identifiers

Chemical Name or Material 1-(2-Cyanophenyl)piperazine
CAS 111373-03-6
Molecular Formula C11H13N3
Formula Weight 187.25
MDL Number MFCD00040793
UN Number UN2922
Beilstein 785973
Synonym 2-(1-Piperazinyl)benzonitrile
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1939206

 
alfa aesar™
L1939206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1939203

 
alfa aesar™
L1939203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1939214

 
alfa aesar™
L1939214
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 187.25
Chemical Name or Material 1-(2-Cyanophenyl)piperazine
Assay 97%
Molecular Formula C11H13N3
MDL Number MFCD00040793
CAS 111373-03-6
Synonym 2-(1-Piperazinyl)benzonitrile
UN Number UN2922
Beilstein 785973
Health Hazard 2 GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
Flash Point >100°C (212°F)
Boiling Point 314° to 315°C
Quantity 5g
Health Hazard 1 Danger
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.