Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(2-Chlorophenyl)imidazole, 99%

$80.76 - $310.39

Chemical Identifiers

Chemical Name or Material 1-(2-Chlorophenyl)imidazole
CAS 51581-50-1
Molecular Formula C9H7ClN2
Formula Weight 178.62
MDL Number MFCD00041205
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0984903

  SDS
alfa aesar™
L0984903
1g Each for $80.76

AAL0984906

  SDS
alfa aesar™
L0984906
5g Each for $310.39
Description & Specifications

Specifications

Formula Weight 178.62
MDL Number MFCD00041205
Chemical Name or Material 1-(2-Chlorophenyl)imidazole
Molecular Formula C9H7ClN2
Assay 99%
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 1 Warning
Boiling Point 108°C (1mmHg)
Quantity 1g
CAS 51581-50-1
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.