Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(2-Chlorophenyl)biguanide hydrochloride, 97%

$48.49 - $177.01

Chemical Identifiers

Chemical Name or Material 1-(2-Chlorophenyl)biguanide hydrochloride
CAS 19579-44-3
Molecular Formula C8H10ClN5·HCl
Formula Weight 248.12
MDL Number MFCD00053016
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2019803

  SDS
alfa aesar™
B2019803
1g Each for $48.49

AAB2019806

  SDS
alfa aesar™
B2019806
5g Each for $177.01
Description & Specifications

Specifications

Formula Weight 248.12
Chemical Name or Material 1-(2-Chlorophenyl)biguanide hydrochloride
Assay 97%
Molecular Formula C8H10ClN5·HCl
MDL Number MFCD00053016
CAS 19579-44-3
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number DU1780100
Melting Point 231° to 234°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.