missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-(2-Chlorophenyl)-3-[4-(trifluoromethoxy)phenyl]urea, 97%, Alfa Aesar™

Chemical Identifiers

CAS 406930-04-9
Molecular Formula C14H10ClF3N2O2
Molecular Weight (g/mol) 330.691
MDL Number MFCD02632826
InChI Key IBVFZYNARZAJSE-UHFFFAOYSA-N
Synonym 1-2-chlorophenyl-3-4-trifluoromethoxy phenyl urea, 3-2-chlorophenyl-1-4-trifluoromethoxy phenyl urea, 1-2-chlorophenyl-3-4-trifluoromethoxyphenyl urea, 1-2-chlorophenyl-3-4-trifluoromethoxy phenyl urea
PubChem CID 951288
IUPAC Name 1-(2-chlorophenyl)-3-[4-(trifluoromethoxy)phenyl]urea
SMILES C1=CC=C(C(=C1)NC(=O)NC2=CC=C(C=C2)OC(F)(F)F)Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH64418MD View Documents Alfa Aesar
H64418MD
250mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH6441803 View Documents Alfa Aesar
H6441803
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH6441806 View Documents Alfa Aesar
H6441806
5g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

406930-04-9
330.691
IBVFZYNARZAJSE-UHFFFAOYSA-N
951288
C1=CC=C(C(=C1)NC(=O)NC2=CC=C(C=C2)OC(F)(F)F)Cl
C14H10ClF3N2O2
MFCD02632826
1-2-chlorophenyl-3-4-trifluoromethoxy phenyl urea, 3-2-chlorophenyl-1-4-trifluoromethoxy phenyl urea, 1-2-chlorophenyl-3-4-trifluoromethoxyphenyl urea, 1-2-chlorophenyl-3-4-trifluoromethoxy phenyl urea
1-(2-chlorophenyl)-3-[4-(trifluoromethoxy)phenyl]urea

Specifications

White
406930-04-9
MFCD02632826
IBVFZYNARZAJSE-UHFFFAOYSA-N
1-(2-chlorophenyl)-3-[4-(trifluoromethoxy)phenyl]urea
951288
Solid
1-(2-Chlorophenyl)-3-[4-(trifluoromethoxy)phenyl]urea
250mg
C14H10ClF3N2O2
1-2-chlorophenyl-3-4-trifluoromethoxy phenyl urea, 3-2-chlorophenyl-1-4-trifluoromethoxy phenyl urea, 1-2-chlorophenyl-3-4-trifluoromethoxyphenyl urea, 1-2-chlorophenyl-3-4-trifluoromethoxy phenyl urea
C1=CC=C(C(=C1)NC(=O)NC2=CC=C(C=C2)OC(F)(F)F)Cl
330.691
330.69
97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning