Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(2-Chloronicotinoyl)piperidine, 97%

$91.91 - $317.09

Chemical Identifiers

Chemical Name or Material 1-(2-Chloronicotinoyl)piperidine
CAS 56149-33-8
Molecular Formula C11H13ClN2O
Formula Weight 224.69
MDL Number MFCD03387556
Synonym 2-Chloro-3-(piperidin-1-ylcarbonyl)pyridine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5084103

 
alfa aesar™
H5084103
1g Each for $317.09

AAH50841MD

 
alfa aesar™
H50841MD
250mg Each for $91.91
Description & Specifications

Specifications

Formula Weight 224.69
Chemical Name or Material 1-(2-Chloronicotinoyl)piperidine
Assay 97%
Molecular Formula C11H13ClN2O
MDL Number MFCD03387556
CAS 56149-33-8
Synonym 2-Chloro-3-(piperidin-1-ylcarbonyl)pyridine
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 61° to 62°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.