Learn More
1-(2-Chloroethyl)piperidine hydrochloride, 98%, Thermo Scientific™
Chemical Identifiers
CAS | 2008-75-5 |
---|---|
Molecular Formula | C7H14ClN·HCl |
Molecular Weight (g/mol) | 184.11 |
MDL Number | MFCD00012837 |
InChI Key | VFLQQZCRHPIGJU-UHFFFAOYSA-N |
Synonym | 1-2-chloroethyl piperidine hydrochloride, 2-piperidinoethylchloride hydrochloride, 2-chloroethylpiperidine hydrochloride, n-chloroethyl piperidine hydrochloride, piperidine, 1-2-chloroethyl-, hydrochloride, 2-piperidinoethyl chloride hydrochloride, piperidinoethyl chloride, hydrochloride, beta-chloroethylpiperidine hydrochloride, 1-2-chloroethyl piperidinium chloride, n-2-chloroethyl piperidine hydrochloride |
PubChem CID | 74826 |
IUPAC Name | 1-(2-chloroethyl)piperidine;hydrochloride |
SMILES | C1CCN(CC1)CCCl.Cl |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC109211000
|
Thermo Scientific Chemicals
109211000 |
100g | Plastic bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC109215000
|
Thermo Scientific Chemicals
109215000 |
500g | Plastic bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
2008-75-5 | |
184.11 | |
VFLQQZCRHPIGJU-UHFFFAOYSA-N | |
74826 | |
C1CCN(CC1)CCCl.Cl |
C7H14ClN·HCl | |
MFCD00012837 | |
1-2-chloroethyl piperidine hydrochloride, 2-piperidinoethylchloride hydrochloride, 2-chloroethylpiperidine hydrochloride, n-chloroethyl piperidine hydrochloride, piperidine, 1-2-chloroethyl-, hydrochloride, 2-piperidinoethyl chloride hydrochloride, piperidinoethyl chloride, hydrochloride, beta-chloroethylpiperidine hydrochloride, 1-2-chloroethyl piperidinium chloride, n-2-chloroethyl piperidine hydrochloride | |
1-(2-chloroethyl)piperidine;hydrochloride |
Specifications
2008-75-5 | |
100.0 | |
Authentic | |
Plastic bottle | |
MFCD00012837 | |
20, 17 | |
Solubility in water: soluble. Other solubilities: soluble in hot ethanol and hot acetone,fairly soluble in cold ethanol,sparingly soluble in cold acetone,insoluble in benzene and toluene | |
C1CCN(CC1)CCCl.Cl | |
184.11 | |
184.11 | |
1-(2-Chloroethyl)piperidine hydrochloride, 98% |
97.5 | |
1% max. (105°C, 3 hrs) | |
98% | |
C7H14ClN·HCl | |
100g | |
1-2-chloroethyl piperidine hydrochloride, 2-piperidinoethylchloride hydrochloride, 2-chloroethylpiperidine hydrochloride, n-chloroethyl piperidine hydrochloride, piperidine, 1-2-chloroethyl-, hydrochloride, 2-piperidinoethyl chloride hydrochloride, piperidinoethyl chloride, hydrochloride, beta-chloroethylpiperidine hydrochloride, 1-2-chloroethyl piperidinium chloride, n-2-chloroethyl piperidine hydrochloride | |
VFLQQZCRHPIGJU-UHFFFAOYSA-N | |
1-(2-chloroethyl)piperidine;hydrochloride | |
74826 | |
98% |
Safety and Handling
GHS H Statement
Fatal if swallowed.
Causes severe skin burns and eye damage.
Suspected of causing genetic defects.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wash face,ha
GHS Signal Word: Danger
EINECSNumber : 217-920-5
RTECSNumber : TM6482500
TSCA : TSCA
RUO â Research Use Only