missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-(2-Chloroethyl)piperidine hydrochloride, 98%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 109211000
Specifications
1-(2-Chloroethyl)piperidine hydrochloride | |
1% max. (105°C, 3 hrs) | |
Beige to White | |
100g | |
2008-75-5 | |
100.0 | |
C7H14ClN·HCl | |
20, 17 | |
Solubility in water: soluble. Other solubilities: soluble in hot ethanol and hot acetone,fairly soluble in cold ethanol,sparingly soluble in cold acetone,insoluble in benzene and toluene | |
C1CCN(CC1)CCCl.Cl | |
184.11 | |
184.11 | |
98% |
Authentic | |
Plastic bottle | |
228°C to 234°C | |
98% | |
97.5 | |
98% | |
MFCD00012837 | |
1-2-chloroethyl piperidine hydrochloride, 1-2-chloroethyl piperidinium chloride, 2-chloroethylpiperidine hydrochloride, 2-piperidinoethyl chloride hydrochloride, 2-piperidinoethylchloride hydrochloride, beta-chloroethylpiperidine hydrochloride, n-2-chloroethyl piperidine hydrochloride, n-chloroethyl piperidine hydrochloride, piperidine, 1-2-chloroethyl-, hydrochloride, piperidinoethyl chloride, hydrochloride | |
VFLQQZCRHPIGJU-UHFFFAOYSA-N | |
1-(2-chloroethyl)piperidine;hydrochloride | |
74826 | |
Powder or Crystals |
Chemical Identifiers
2008-75-5 | |
184.11 | |
VFLQQZCRHPIGJU-UHFFFAOYSA-N | |
74826 | |
C1CCN(CC1)CCCl.Cl |
C7H14ClN·HCl | |
MFCD00012837 | |
1-2-chloroethyl piperidine hydrochloride, 1-2-chloroethyl piperidinium chloride, 2-chloroethylpiperidine hydrochloride, 2-piperidinoethyl chloride hydrochloride, 2-piperidinoethylchloride hydrochloride, beta-chloroethylpiperidine hydrochloride, n-2-chloroethyl piperidine hydrochloride, n-chloroethyl piperidine hydrochloride, piperidine, 1-2-chloroethyl-, hydrochloride, piperidinoethyl chloride, hydrochloride | |
1-(2-chloroethyl)piperidine;hydrochloride |
Safety and Handling
GHS H Statement
Fatal if swallowed.
Causes severe skin burns and eye damage.
Suspected of causing genetic defects.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wash face,ha
GHS Signal Word: Danger
EINECSNumber : 217-920-5
RTECSNumber : TM6482500
TSCA : TSCA