missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-(2-Chloroethyl)-4-methoxybenzene, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 123111000

 View more versions of this product

Catalog No. AC123111000



Description

Description

Specifications

Specifications

1-(2-Chloroethyl)-4-methoxybenzene
122°C
Glass bottle
1.13
Colorless to Yellow
97%
96.0
96% min. (GC)
CH3OC6H4(CH2)2Cl
1-2-chloroethyl-4-methoxybenzene, 2-4-methoxyphenyl ethylchloride, 2-4-methoxyphenyl-ethylchloride, 4-2-chloroethyl anisole, 4-2-chloroethyl phenyl methyl ether, 4-methoxyphenethyl chloride, 4-methoxyphenylethyl chloride, acmc-1c78x, benzene, 1-2-chloroethyl-4-methoxy, p-2-chloro ethyl anisole
PMIAMRAWHYEPNH-UHFFFAOYSA-N
1-(2-chloroethyl)-4-methoxybenzene
87513
Liquid
1.1300g/mL
Authentic
1.5360 to 1.538
125°C to 128°C (13.0 mbar)
100mL
18217-00-0
100.0
C9H11ClO
MFCD00044718
Solubility in water: 130mg/l (20°C). Other solubilities: soluble in methanol,ethanol,2-propanol,,n,n-dimethylformamide,acetone and dichloromethane
COC1=CC=C(CCCl)C=C1
170.64
170.64
97%

Chemical Identifiers

18217-00-0
170.64
PMIAMRAWHYEPNH-UHFFFAOYSA-N
87513
COC1=CC=C(CCCl)C=C1
C9H11ClO
MFCD00044718
1-2-chloroethyl-4-methoxybenzene, 2-4-methoxyphenyl ethylchloride, 2-4-methoxyphenyl-ethylchloride, 4-2-chloroethyl anisole, 4-2-chloroethyl phenyl methyl ether, 4-methoxyphenethyl chloride, 4-methoxyphenylethyl chloride, acmc-1c78x, benzene, 1-2-chloroethyl-4-methoxy, p-2-chloro ethyl anisole
1-(2-chloroethyl)-4-methoxybenzene
Safety and Handling

Safety and Handling

GHS H Statement:
Suspected of causing genetic defects.
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 242-099-5

TSCA : TSCA

SDS Documents