missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-(2-Chloroethyl)-4-methoxybenzene, 97%, ACROS Organics™

$219.67 - $219.67

Chemical Identifiers

CAS 18217-00-0
Molecular Formula C9H11ClO
Molecular Weight (g/mol) 170.64
MDL Number MFCD00044718
InChI Key PMIAMRAWHYEPNH-UHFFFAOYSA-N
Synonym 1-2-chloroethyl-4-methoxybenzene, 2-4-methoxyphenyl ethylchloride, 2-4-methoxyphenyl-ethylchloride, 4-2-chloroethyl anisole, 4-2-chloroethyl phenyl methyl ether, 4-methoxyphenethyl chloride, 4-methoxyphenylethyl chloride, acmc-1c78x, benzene, 1-2-chloroethyl-4-methoxy, p-2-chloro ethyl anisole
PubChem CID 87513
IUPAC Name 1-(2-chloroethyl)-4-methoxybenzene
SMILES COC1=CC=C(CCCl)C=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC123110050
SDS View Documents
Thermo Fisher Scientific
123110050
5mL Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC123111000
SDS View Documents
Thermo Fisher Scientific
123111000
100mL Glass bottle
Each for $219.67
 
Description

Description

Specifications

Chemical Identifiers

18217-00-0
170.64
PMIAMRAWHYEPNH-UHFFFAOYSA-N
87513
COC1=CC=C(CCCl)C=C1
C9H11ClO
MFCD00044718
1-2-chloroethyl-4-methoxybenzene, 2-4-methoxyphenyl ethylchloride, 2-4-methoxyphenyl-ethylchloride, 4-2-chloroethyl anisole, 4-2-chloroethyl phenyl methyl ether, 4-methoxyphenethyl chloride, 4-methoxyphenylethyl chloride, acmc-1c78x, benzene, 1-2-chloroethyl-4-methoxy, p-2-chloro ethyl anisole
1-(2-chloroethyl)-4-methoxybenzene

Specifications

1.1300g/mL
Authentic
1.5360 to 1.538
125°C to 128°C (13.0 mbar)
5mL
96.0
96% min. (GC)
CH3OC6H4(CH2)2Cl
1-2-chloroethyl-4-methoxybenzene, 2-4-methoxyphenyl ethylchloride, 2-4-methoxyphenyl-ethylchloride, 4-2-chloroethyl anisole, 4-2-chloroethyl phenyl methyl ether, 4-methoxyphenethyl chloride, 4-methoxyphenylethyl chloride, acmc-1c78x, benzene, 1-2-chloroethyl-4-methoxy, p-2-chloro ethyl anisole
PMIAMRAWHYEPNH-UHFFFAOYSA-N
1-(2-chloroethyl)-4-methoxybenzene
87513
Liquid
1-(2-Chloroethyl)-4-methoxybenzene, 97%
122°C
Glass bottle
1.13
Colorless to Yellow
18217-00-0
100.0
C9H11ClO
MFCD00044718
Solubility in water: 130mg/l (20°C). Other solubilities: soluble in methanol,ethanol,2-propanol,,n,n-dimethylformamide,acetone and dichloromethane
COC1=CC=C(CCCl)C=C1
170.64
170.64
97%
Safety and Handling

Safety and Handling

GHS H Statement:
Suspected of causing genetic defects.
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 242-099-5

TSCA : TSCA

SDS
Product Certifications