missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(2-Chloroethyl)-4-fluorobenzene, 97%

$117.05 - $450.30

Chemical Identifiers

CAS 332-43-4
Molecular Formula C8H8ClF
Molecular Weight (g/mol) 158.6
MDL Number MFCD00155000
InChI Key LPUURRVXQCVXCX-UHFFFAOYSA-N
Synonym 1-2-chloroethyl-4-fluorobenzene, 4-fluorophenethyl chloride, 2-4-fluorophenyl ethyl chloride, 1-2'-chloroethyl-4-fluorobenzene, 1-2-chloro-ethyl-4-fluoro-benzene, benzene, 1-2-chloroethyl-4-fluoro, acmc-1anql, 4-fluorophenetyl chloride, ksc570e7d, 1-fluoro-4-2-chloroethyl benzene
PubChem CID 67622
IUPAC Name 1-(2-chloroethyl)-4-fluorobenzene
SMILES C1=CC(=CC=C1CCCl)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1671903 View Documents Alfa Aesar™
L1671903
1g Each for $117.05
 
AAL1671906 missing translation for 'viewDocuments' Alfa Aesar™
L1671906
5g Each for $450.30
 
Specifications

Chemical Identifiers

332-43-4
158.6
LPUURRVXQCVXCX-UHFFFAOYSA-N
67622
C1=CC(=CC=C1CCCl)F
C8H8ClF
MFCD00155000
1-2-chloroethyl-4-fluorobenzene, 4-fluorophenethyl chloride, 2-4-fluorophenyl ethyl chloride, 1-2'-chloroethyl-4-fluorobenzene, 1-2-chloro-ethyl-4-fluoro-benzene, benzene, 1-2-chloroethyl-4-fluoro, acmc-1anql, 4-fluorophenetyl chloride, ksc570e7d, 1-fluoro-4-2-chloroethyl benzene
1-(2-chloroethyl)-4-fluorobenzene

Specifications

98°C to 99°C (26mmHg)
332-43-4
MFCD00155000
1-2-chloroethyl-4-fluorobenzene, 4-fluorophenethyl chloride, 2-4-fluorophenyl ethyl chloride, 1-2'-chloroethyl-4-fluorobenzene, 1-2-chloro-ethyl-4-fluoro-benzene, benzene, 1-2-chloroethyl-4-fluoro, acmc-1anql, 4-fluorophenetyl chloride, ksc570e7d, 1-fluoro-4-2-chloroethyl benzene
C1=CC(=CC=C1CCCl)F
158.6
158.6
1-(2-Chloroethyl)-4-fluorobenzene
1g
C8H8ClF
2079481
LPUURRVXQCVXCX-UHFFFAOYSA-N
1-(2-chloroethyl)-4-fluorobenzene
67622
97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

Warning