Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(2-Chloro-6-nitrophenyl)pyrrolidine, 97%

Chemical Identifiers

Chemical Name or Material 1-(2-Chloro-6-nitrophenyl)pyrrolidine
CAS 59504-30-2
Formula Weight 226.66
Beilstein 4317456
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6309403

 
alfa aesar™
H6309403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6309406

 
alfa aesar™
H6309406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH63094MD

 
alfa aesar™
H63094MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 226.66
Physical Form Solid
CAS 59504-30-2
Beilstein 4317456
Chemical Name or Material 1-(2-Chloro-6-nitrophenyl)pyrrolidine
Quantity 1g
Assay 97%