Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(2-Chloro-6-nitrophenyl)piperidine, 97%

Chemical Identifiers

Chemical Name or Material 1-(2-Chloro-6-nitrophenyl)piperidine
CAS 3970-42-1
Formula Weight 240.69
Beilstein 1317515
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6328303

 
alfa aesar™
H6328303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6328306

 
alfa aesar™
H6328306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH63283MD

 
alfa aesar™
H63283MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 240.69
Chemical Name or Material 1-(2-Chloro-6-nitrophenyl)piperidine
Physical Form Solid
Assay 97%
CAS 3970-42-1
Beilstein 1317515
Quantity 1g