missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(2-Chloro-6-nitrophenyl)piperazine, 97%

$106.26 - $1,406.42

Chemical Identifiers

CAS 332023-12-8
Molecular Formula C10H12ClN3O2
Molecular Weight (g/mol) 241.675
MDL Number MFCD01173865
InChI Key GKOXZZHKOZLHRQ-UHFFFAOYSA-N
Synonym 1-2-chloro-6-nitrophenyl piperazine, 1-2-chloro-6-nitro-phenyl-piperazine, piperazine, 1-2-chloro-6-nitrophenyl, cambridge id 5749393, tos-bb-0566, 2-chloro-6-nitrophenyl piperazine, 6-chloro-2-nitrophenyl piperazine, 1-2-chloro-6-nitrophenyl-piperazine, 3-chloro-2-piperazin-1-yl nitrobenzene
PubChem CID 2771408
IUPAC Name 1-(2-chloro-6-nitrophenyl)piperazine
SMILES C1CN(CCN1)C2=C(C=CC=C2Cl)[N+](=O)[O-]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH63461MD View Documents Alfa Aesar™
H63461MD
250mg Each for $106.26
 
AAH6346103 View Documents Alfa Aesar™
H6346103
1g Each for $337.73
 
AAH6346106 View Documents Alfa Aesar™
H6346106
5g Each for $1,406.42
 
Specifications

Chemical Identifiers

332023-12-8
241.675
GKOXZZHKOZLHRQ-UHFFFAOYSA-N
2771408
C1CN(CCN1)C2=C(C=CC=C2Cl)[N+](=O)[O-]
C10H12ClN3O2
MFCD01173865
1-2-chloro-6-nitrophenyl piperazine, 1-2-chloro-6-nitro-phenyl-piperazine, piperazine, 1-2-chloro-6-nitrophenyl, cambridge id 5749393, tos-bb-0566, 2-chloro-6-nitrophenyl piperazine, 6-chloro-2-nitrophenyl piperazine, 1-2-chloro-6-nitrophenyl-piperazine, 3-chloro-2-piperazin-1-yl nitrobenzene
1-(2-chloro-6-nitrophenyl)piperazine

Specifications

250mg
C10H12ClN3O2
1-2-chloro-6-nitrophenyl piperazine, 1-2-chloro-6-nitro-phenyl-piperazine, piperazine, 1-2-chloro-6-nitrophenyl, cambridge id 5749393, tos-bb-0566, 2-chloro-6-nitrophenyl piperazine, 6-chloro-2-nitrophenyl piperazine, 1-2-chloro-6-nitrophenyl-piperazine, 3-chloro-2-piperazin-1-yl nitrobenzene
C1CN(CCN1)C2=C(C=CC=C2Cl)[N+](=O)[O-]
241.675
241.67
97%
332023-12-8
MFCD01173865
GKOXZZHKOZLHRQ-UHFFFAOYSA-N
1-(2-chloro-6-nitrophenyl)piperazine
2771408
Solid
1-(2-Chloro-6-nitrophenyl)piperazine