missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(2-Chloro-6-nitrophenyl)piperazine, 97%

Manufacturer:  Alfa Aesar™ H6346106

 View more versions of this product

Catalog No. AAH6346106



Specifications

Specifications

1-(2-Chloro-6-nitrophenyl)piperazine
332023-12-8
MFCD01173865
GKOXZZHKOZLHRQ-UHFFFAOYSA-N
1-(2-chloro-6-nitrophenyl)piperazine
2771408
Solid
5g
C10H12ClN3O2
1-2-chloro-6-nitrophenyl piperazine, 1-2-chloro-6-nitro-phenyl-piperazine, piperazine, 1-2-chloro-6-nitrophenyl, cambridge id 5749393, tos-bb-0566, 2-chloro-6-nitrophenyl piperazine, 6-chloro-2-nitrophenyl piperazine, 1-2-chloro-6-nitrophenyl-piperazine, 3-chloro-2-piperazin-1-yl nitrobenzene
C1CN(CCN1)C2=C(C=CC=C2Cl)[N+](=O)[O-]
241.675
241.67
97%

Chemical Identifiers

332023-12-8
241.675
GKOXZZHKOZLHRQ-UHFFFAOYSA-N
2771408
C1CN(CCN1)C2=C(C=CC=C2Cl)[N+](=O)[O-]
C10H12ClN3O2
MFCD01173865
1-2-chloro-6-nitrophenyl piperazine, 1-2-chloro-6-nitro-phenyl-piperazine, piperazine, 1-2-chloro-6-nitrophenyl, cambridge id 5749393, tos-bb-0566, 2-chloro-6-nitrophenyl piperazine, 6-chloro-2-nitrophenyl piperazine, 1-2-chloro-6-nitrophenyl-piperazine, 3-chloro-2-piperazin-1-yl nitrobenzene
1-(2-chloro-6-nitrophenyl)piperazine
Documents