missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(2-Chloro-6-nitrophenyl)azepane, 97%

Chemical Identifiers

Formula Weight 254.71
UN Number UN3259
Synonym 1-(2-Chloro-6-nitrophenyl)hexamethyleneimine
Chemical Name or Material 1-(2-Chloro-6-nitrophenyl)azepane
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6384703 SDS Alfa Aesar™
H6384703
1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH6384706 SDS Alfa Aesar™
H6384706
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH63847MD SDS Alfa Aesar™
H63847MD
250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

254.71
UN3259
1-(2-Chloro-6-nitrophenyl)hexamethyleneimine
1-(2-Chloro-6-nitrophenyl)azepane

Specifications

1g
254.71
1-(2-Chloro-6-nitrophenyl)hexamethyleneimine
97%
Solid
UN3259
1-(2-Chloro-6-nitrophenyl)azepane

Safety and Handling

GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.