missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(2-Chloro-6-nitrophenyl)-4-methylpiperazine, 97%

Chemical Identifiers

CAS 189761-96-4
Molecular Formula C11H14ClN3O2
Molecular Weight (g/mol) 255.702
InChI Key SKTYGQGYDUZEPT-UHFFFAOYSA-N
Synonym 1-2-chloro-6-nitrophenyl-4-methylpiperazine, cbmicro_045682, 1-6-chloro-2-nitrophenyl-4-methylpiperazine
PubChem CID 2885663
IUPAC Name 1-(2-chloro-6-nitrophenyl)-4-methylpiperazine
SMILES CN1CCN(CC1)C2=C(C=CC=C2Cl)[N+](=O)[O-]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH63437MD View Documents Alfa Aesar™
H63437MD
250mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH6343703 View Documents Alfa Aesar™
H6343703
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH6343706 View Documents Alfa Aesar™
H6343706
5g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

189761-96-4
255.702
1-2-chloro-6-nitrophenyl-4-methylpiperazine, cbmicro_045682, 1-6-chloro-2-nitrophenyl-4-methylpiperazine
1-(2-chloro-6-nitrophenyl)-4-methylpiperazine
C11H14ClN3O2
SKTYGQGYDUZEPT-UHFFFAOYSA-N
2885663
CN1CCN(CC1)C2=C(C=CC=C2Cl)[N+](=O)[O-]

Specifications

250mg
C11H14ClN3O2
SKTYGQGYDUZEPT-UHFFFAOYSA-N
1-(2-chloro-6-nitrophenyl)-4-methylpiperazine
2885663
Solid
1-(2-Chloro-6-nitrophenyl)-4-methylpiperazine
189761-96-4
1-2-chloro-6-nitrophenyl-4-methylpiperazine, cbmicro_045682, 1-6-chloro-2-nitrophenyl-4-methylpiperazine
CN1CCN(CC1)C2=C(C=CC=C2Cl)[N+](=O)[O-]
255.702
255.7
97%