missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(2-Chloro-6-nitrophenyl)-4-ethylpiperazine, 97%

Chemical Identifiers

Formula Weight 269.73
Chemical Name or Material 1-(2-Chloro-6-nitrophenyl)-4-ethylpiperazine
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6351303 Alfa Aesar™
H6351303
1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH6351306 Alfa Aesar™
H6351306
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH63513MD Alfa Aesar™
H63513MD
250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

269.73
1-(2-Chloro-6-nitrophenyl)-4-ethylpiperazine

Specifications

1g
269.73
97%
Solid
1-(2-Chloro-6-nitrophenyl)-4-ethylpiperazine