missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(2-Chloro-4-nitrophenyl)pyrrolidine, 97%

Manufacturer:  Alfa Aesar™ H6341503

 View more versions of this product

Catalog No. AAH6341503



Specifications

Specifications

1-(2-Chloro-4-nitrophenyl)pyrrolidine
55403-91-3
1-2-chloro-4-nitrophenyl pyrrolidine, 4-pyrrolidino-3-chloronitrobenzene, pyrrolidine, 1-2-chloro-4-nitrophenyl, 2-chloro-4-nitrophenyl pyrrolidine, cambridge id 5139302, pyrrolidine, 1-2-chloro-4-nitrophenyl
C1CCN(C1)C2=C(C=C(C=C2)[N+](=O)[O-])Cl
226.66
226.66
97%
1g
C10H11ClN2O2
XMLGZIMCJDYHOD-UHFFFAOYSA-N
1-(2-chloro-4-nitrophenyl)pyrrolidine
108707
Solid

Chemical Identifiers

55403-91-3
226.66
1-2-chloro-4-nitrophenyl pyrrolidine, 4-pyrrolidino-3-chloronitrobenzene, pyrrolidine, 1-2-chloro-4-nitrophenyl, 2-chloro-4-nitrophenyl pyrrolidine, cambridge id 5139302, pyrrolidine, 1-2-chloro-4-nitrophenyl
1-(2-chloro-4-nitrophenyl)pyrrolidine
C10H11ClN2O2
XMLGZIMCJDYHOD-UHFFFAOYSA-N
108707
C1CCN(C1)C2=C(C=C(C=C2)[N+](=O)[O-])Cl
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents