missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(2-Chloro-4-nitrophenyl)pyrrolidine, 97%

$35.93 - $422.54

Chemical Identifiers

CAS 55403-91-3
Molecular Formula C10H11ClN2O2
Molecular Weight (g/mol) 226.66
InChI Key XMLGZIMCJDYHOD-UHFFFAOYSA-N
Synonym 1-2-chloro-4-nitrophenyl pyrrolidine, 4-pyrrolidino-3-chloronitrobenzene, pyrrolidine, 1-2-chloro-4-nitrophenyl, 2-chloro-4-nitrophenyl pyrrolidine, cambridge id 5139302, pyrrolidine, 1-2-chloro-4-nitrophenyl
PubChem CID 108707
IUPAC Name 1-(2-chloro-4-nitrophenyl)pyrrolidine
SMILES C1CCN(C1)C2=C(C=C(C=C2)[N+](=O)[O-])Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH63415MD View Documents Alfa Aesar™
H63415MD
250mg Each for $35.93
 
AAH6341503 View Documents Alfa Aesar™
H6341503
1g Each for $106.26
 
AAH6341506 View Documents Alfa Aesar™
H6341506
5g Each for $422.54
 
Specifications

Chemical Identifiers

55403-91-3
226.66
1-2-chloro-4-nitrophenyl pyrrolidine, 4-pyrrolidino-3-chloronitrobenzene, pyrrolidine, 1-2-chloro-4-nitrophenyl, 2-chloro-4-nitrophenyl pyrrolidine, cambridge id 5139302, pyrrolidine, 1-2-chloro-4-nitrophenyl
1-(2-chloro-4-nitrophenyl)pyrrolidine
C10H11ClN2O2
XMLGZIMCJDYHOD-UHFFFAOYSA-N
108707
C1CCN(C1)C2=C(C=C(C=C2)[N+](=O)[O-])Cl

Specifications

250mg
C10H11ClN2O2
XMLGZIMCJDYHOD-UHFFFAOYSA-N
1-(2-chloro-4-nitrophenyl)pyrrolidine
108707
Solid
1-(2-Chloro-4-nitrophenyl)pyrrolidine
55403-91-3
1-2-chloro-4-nitrophenyl pyrrolidine, 4-pyrrolidino-3-chloronitrobenzene, pyrrolidine, 1-2-chloro-4-nitrophenyl, 2-chloro-4-nitrophenyl pyrrolidine, cambridge id 5139302, pyrrolidine, 1-2-chloro-4-nitrophenyl
C1CCN(C1)C2=C(C=C(C=C2)[N+](=O)[O-])Cl
226.66
226.66
97%
Safety and Handling

Safety and Handling

TSCA : TSCA