missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(2-Chloro-4-nitrophenyl)piperidine, 97%

$28.84 - $337.73

Chemical Identifiers

CAS 55403-25-3
Molecular Formula C11H13ClN2O2
Molecular Weight (g/mol) 240.687
InChI Key AIZKHORWEJERNY-UHFFFAOYSA-N
Synonym 1-2-chloro-4-nitrophenyl piperidine, 1-2-chloro-4-nitro-phenyl-piperidine, 2-chloro-4-nitrophenyl piperidine, maybridge1_000868, piperidine, 1-2-chloro-4-nitrophenyl
PubChem CID 2757742
IUPAC Name 1-(2-chloro-4-nitrophenyl)piperidine
SMILES C1CCN(CC1)C2=C(C=C(C=C2)[N+](=O)[O-])Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH63972MD View Documents Alfa Aesar™
H63972MD
250mg Each for $28.84
 
AAH6397203 View Documents Alfa Aesar™
H6397203
1g Each for $85.14
 
AAH6397206 View Documents Alfa Aesar™
H6397206
5g Each for $337.73
 
Specifications

Chemical Identifiers

55403-25-3
240.687
1-2-chloro-4-nitrophenyl piperidine, 1-2-chloro-4-nitro-phenyl-piperidine, 2-chloro-4-nitrophenyl piperidine, maybridge1_000868, piperidine, 1-2-chloro-4-nitrophenyl
1-(2-chloro-4-nitrophenyl)piperidine
C11H13ClN2O2
AIZKHORWEJERNY-UHFFFAOYSA-N
2757742
C1CCN(CC1)C2=C(C=C(C=C2)[N+](=O)[O-])Cl

Specifications

250mg
C11H13ClN2O2
AIZKHORWEJERNY-UHFFFAOYSA-N
1-(2-chloro-4-nitrophenyl)piperidine
2757742
Solid
1-(2-Chloro-4-nitrophenyl)piperidine
55403-25-3
1-2-chloro-4-nitrophenyl piperidine, 1-2-chloro-4-nitro-phenyl-piperidine, 2-chloro-4-nitrophenyl piperidine, maybridge1_000868, piperidine, 1-2-chloro-4-nitrophenyl
C1CCN(CC1)C2=C(C=C(C=C2)[N+](=O)[O-])Cl
240.687
240.69
97%