missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(2-Chloro-4-nitrophenyl)piperazine, 97%

Manufacturer:  Alfa Aesar™ H6323203

 View more versions of this product

Catalog No. AAH6323203



Specifications

Specifications

1-(2-Chloro-4-nitrophenyl)piperazine
114878-60-3
MFCD00640776
JNUWBYPJOBSXDN-UHFFFAOYSA-N
1-(2-chloro-4-nitrophenyl)piperazine
2771331
Solid
1g
C10H12ClN3O2
1-2-chloro-4-nitrophenyl piperazine, 2-chloro-4-nitrophenyl piperazine, 1-2-chloro-4-nitrophenyl-piperazine, piperazine, 1-2-chloro-4-nitrophenyl, 1-2-chloro-4-nitro-phenyl-piperazine, piperazine,1-2-chloro-4-nitrophenyl, pubchem9343, acmc-1bvwp, cambridge id 5668126, 1-2-chloro-4-nitro-phenyl piperazine
C1CN(CCN1)C2=C(C=C(C=C2)[N+](=O)[O-])Cl
241.675
241.67
97%

Chemical Identifiers

114878-60-3
241.675
JNUWBYPJOBSXDN-UHFFFAOYSA-N
2771331
C1CN(CCN1)C2=C(C=C(C=C2)[N+](=O)[O-])Cl
C10H12ClN3O2
MFCD00640776
1-2-chloro-4-nitrophenyl piperazine, 2-chloro-4-nitrophenyl piperazine, 1-2-chloro-4-nitrophenyl-piperazine, piperazine, 1-2-chloro-4-nitrophenyl, 1-2-chloro-4-nitro-phenyl-piperazine, piperazine,1-2-chloro-4-nitrophenyl, pubchem9343, acmc-1bvwp, cambridge id 5668126, 1-2-chloro-4-nitro-phenyl piperazine
1-(2-chloro-4-nitrophenyl)piperazine
Documents